siRNA Details
Gene Symbol: C8orf74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040032.1 | NP_001035121.1 | 3 | 315 | ||
XM_011543816.2 | XP_011542118.1 | 3 | 416 | |||
GenBank mRNA | BC065837.1 | - | 3 | 315 | ||
BC132852.1 | - | 3 | 315 | 40146874 | ||
BC132854.1 | - | 3 | 315 | 40146876 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
