siRNA Details
Gene Symbol: CYTH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004228.6 | NP_004219.3 | 12 | 4247 | ||
NM_017457.5 | NP_059431.1 | 13 | 4250 | |||
GenBank mRNA | AF318380.1 | - | 2 | 1664 | ||
BC036615.1 | - | 1 | 2117 | |||
BC047631.1 | - | 1 | 2112 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
