siRNA Details
Gene Symbol: SHE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010846.2 | NP_001010846.1 | 2 | 666 | ||
NR_135169.1 | 2 | 666 | ||||
XM_005244891.4 | XP_005244948.1 | 2 | 669 | |||
XM_011509163.2 | XP_011507465.1 | 2 | 675 | |||
XM_017000256.1 | XP_016855745.1 | 2 | 669 | |||
GenBank mRNA | AK074067.1 | - | 2 | 85 | ||
BC117210.1 | - | 2 | 666 | 40125761 | ||
BC143765.1 | - | 2 | 665 | 9052279 | ||
CR936667.1 | - | 2 | 114 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
