siRNA Details
Gene Symbol: SHE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010846.2 | NP_001010846.1 | 6 | 1423 | ||
NR_135169.1 | 6 | 1418 | ||||
XM_011509163.2 | XP_011507465.1 | 6 | 1432 | |||
GenBank mRNA | AK074067.1 | - | 842 | |||
AK126006.1 | - | 4 | 688 | |||
BC117210.1 | - | 6 | 1423 | 40125761 | ||
BC143765.1 | - | 6 | 1422 | 9052279 | ||
CR936667.1 | - | 6 | 866 | |||
CR936736.1 | - | 3 | 2098 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
