siRNA Details
Gene Symbol: SHE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010846.2 | NP_001010846.1 | 6 | 2881 | ||
NR_135169.1 | 6 | 2876 | ||||
GenBank mRNA | AK074067.1 | - | 7 | 2298 | ||
CR936667.1 | - | 6 | 2323 | |||
CR936736.1 | - | 3 | 3555 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
