siRNA Details
Gene Symbol: MUC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164462.1 | NP_001157934.1 | 3 | 15017 | ||
GenBank mRNA | AF147790.1 | - | 2 | 769 | ||
AK024856.1 | - | 2 | 524 | |||
BC127877.1 | - | 2 | 787 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
