siRNA Details
Gene Symbol: ESCO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017420.2 | NP_001017420.1 | 3 | 524 | ||
XM_011544421.2 | XP_011542723.1 | 3 | 577 | |||
XM_011544422.2 | XP_011542724.1 | 3 | 525 | |||
GenBank mRNA | AF306676.1 | - | 2 | 465 | ||
AF306679.1 | - | 3 | 521 | |||
AK124215.1 | - | 3 | 526 | |||
AL832666.1 | - | 3 | 511 | |||
AY882862.1 | - | 3 | 524 | |||
BC017933.1 | - | 3 | 511 | |||
BC034641.1 | - | 3 | 511 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
