siRNA Details
Gene Symbol: ESCO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017420.2 | NP_001017420.1 | 10 | 1742 | ||
XM_011544421.2 | XP_011542723.1 | 10 | 1795 | |||
XM_011544422.2 | XP_011542724.1 | 10 | 1743 | |||
GenBank mRNA | AF306679.1 | - | 10 | 1739 | ||
AK124215.1 | - | 10 | 1744 | |||
AL832666.1 | - | 10 | 1729 | |||
AY882862.1 | - | 10 | 1742 | |||
BC034641.1 | - | 8 | 1576 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
