siRNA Details
Gene Symbol: GUSBP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024448.2 | 2 | 473 | |||
GenBank mRNA | AK056875.1 | - | 2 | 406 | ||
AK091837.1 | - | 2 | 331 | |||
AK300413.1 | - | 2 | 375 | |||
BC063045.1 | - | 2 | 292 | |||
BX538181.1 | - | 2 | 321 | |||
DC322250.1 | - | 2 | 474 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
