siRNA Details
Gene Symbol: PNMA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007257.5 | NP_009188.1 | 3 | 1746 | ||
XM_011544365.2 | XP_011542667.1 | 3 | 1845 | |||
GenBank mRNA | AB020690.1 | - | 1 | 1153 | ||
AF083114.1 | - | 1 | 332 | |||
AF083115.1 | - | 1 | 734 | |||
AK055325.1 | - | 3 | 1688 | |||
BC036489.1 | - | 3 | 1676 | 5262276 | ||
BC047515.1 | - | 3 | 1651 | 5294654 | ||
BC062301.1 | - | 1 | 1227 | 6580976 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
