siRNA Details
Gene Symbol: PNMA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007257.5 | NP_009188.1 | 3 | 3027 | ||
XM_011544365.2 | XP_011542667.1 | 3 | 3126 | |||
GenBank mRNA | AB020690.1 | - | 1 | 2434 | ||
BC036489.1 | - | 3 | 2957 | 5262276 | ||
BC047515.1 | - | 3 | 2934 | 5294654 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
