siRNA Details
Gene Symbol: RPLP0P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002775.2 | 5 | 3251 | |||
GenBank mRNA | AK095332.1 | - | 5 | 3254 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
