siRNA Details
Gene Symbol: ADAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012969.3 | NP_001012987.1 | 5 | 1014 | ||
NM_001159280.2 | NP_001152752.1 | 6 | 1115 | |||
NM_001324364.1 | NP_001311293.1 | 6 | 828 | |||
NM_001324365.1 | NP_001311294.1 | 6 | 979 | |||
NM_001324366.1 | NP_001311295.1 | 6 | 1115 | |||
NM_001324367.1 | NP_001311296.1 | 6 | 1115 | |||
NM_001324368.1 | NP_001311297.1 | 6 | 1115 | |||
XM_011521291.2 | XP_011519593.1 | 5 | 591 | |||
XM_017021964.1 | XP_016877453.1 | 6 | 701 | |||
XM_017021965.1 | XP_016877454.1 | 6 | 680 | |||
XM_017021966.1 | XP_016877455.1 | 5 | 617 | |||
XM_017021967.1 | XP_016877456.1 | 4 | 365 | |||
XM_017021968.1 | XP_016877457.1 | 6 | 824 | |||
XM_017021969.1 | XP_016877458.1 | 5 | 591 | |||
GenBank mRNA | AK298870.1 | - | 6 | 796 | ||
BC075857.1 | - | 5 | 643 | 5199077 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
