siRNA Details
Gene Symbol: ADAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159280.2 | NP_001152752.1 | 11 | 1657 | ||
NM_001324366.1 | NP_001311295.1 | 12 | 1738 | |||
XM_011521291.2 | XP_011519593.1 | 11 | 1214 | |||
XM_017021964.1 | XP_016877453.1 | 12 | 1324 | |||
XM_017021965.1 | XP_016877454.1 | 12 | 1303 | |||
XM_017021966.1 | XP_016877455.1 | 11 | 1240 | |||
XM_017021967.1 | XP_016877456.1 | 10 | 988 | |||
GenBank mRNA | AK126583.1 | - | 6 | 573 | ||
AK298870.1 | - | 11 | 1338 | |||
BX647599.1 | - | 3 | 286 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
