siRNA Details
Gene Symbol: ADAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159280.2 | NP_001152752.1 | 12 | 1797 | ||
NM_001324366.1 | NP_001311295.1 | 13 | 1878 | |||
XM_011521291.2 | XP_011519593.1 | 12 | 1354 | |||
XM_017021964.1 | XP_016877453.1 | 13 | 1464 | |||
XM_017021965.1 | XP_016877454.1 | 13 | 1443 | |||
XM_017021966.1 | XP_016877455.1 | 12 | 1380 | |||
XM_017021967.1 | XP_016877456.1 | 11 | 1128 | |||
XM_017021968.1 | XP_016877457.1 | 12 | 1486 | |||
XM_017021969.1 | XP_016877458.1 | 10 | 1172 | |||
GenBank mRNA | AK126583.1 | - | 7 | 713 | ||
AK298870.1 | - | 12 | 1478 | |||
BX647599.1 | - | 3 | 1134 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
