siRNA Details
Gene Symbol: OR5E1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_027711.1 | 1 | 61 | |||
GenBank mRNA | AF309699.1 | - | LIT2079 OR5E1P. |
1 | 61 | |
X89671.1 | - | 1 | 38 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
