siRNA Details
Gene Symbol: RNF187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010858.2 | NP_001010858.2 | 4 | 1681 | ||
GenBank mRNA | BC000821.1 | - | 1 | 522 | ||
BC008022.1 | - | 2 | 886 | |||
BC012758.2 | - | 4 | 1655 | |||
BC053571.1 | - | 1 | 494 | |||
BC064481.1 | - | 4 | 1215 | |||
BC080645.1 | - | 4 | 1215 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
