siRNA Details
Gene Symbol: SYNE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015180.4 | NP_055995.4 | 29, 30 | 4023 | ||
NM_182914.2 | NP_878918.2 | 29, 30 | 4023 | |||
XM_005267454.1 | XP_005267511.1 | 29, 30 | 4023 | |||
XM_005267456.1 | XP_005267513.1 | 29, 30 | 4023 | |||
XM_005267457.1 | XP_005267514.1 | 29, 30 | 4023 | |||
XM_005267458.1 | XP_005267515.1 | 29, 30 | 4023 | |||
XM_005267459.1 | XP_005267516.1 | 29, 30 | 4023 | |||
XM_011536574.1 | XP_011534876.1 | 29, 30 | 4023 | |||
XM_011536575.2 | XP_011534877.1 | 29, 30 | 4000 | |||
XM_011536576.2 | XP_011534878.1 | 30, 31 | 4180 | |||
XM_011536577.2 | XP_011534879.1 | 29, 30 | 3947 | |||
XM_011536578.1 | XP_011534880.1 | 29, 30 | 4023 | |||
XM_011536579.1 | XP_011534881.1 | 29, 30 | 4023 | |||
XM_011536580.1 | XP_011534882.1 | 29, 30 | 4023 | |||
XM_011536581.1 | XP_011534883.1 | 29, 30 | 4023 | |||
XM_011536582.1 | XP_011534884.1 | 29, 30 | 4023 | |||
XM_011536584.2 | XP_011534886.1 | 29, 30 | 4023 | |||
XM_017021101.1 | XP_016876590.1 | 29, 30 | 4025 | |||
XM_017021102.1 | XP_016876591.1 | 28, 29 | 4068 | |||
GenBank mRNA | AF357236.1 | - | 28, 29 | 3793 | ||
AF435011.1 | - | 29, 30 | 4005 | |||
AF495911.1 | - | 29, 30 | 3990 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
