siRNA Details
Gene Symbol: C2orf68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013649.3 | NP_001013671.2 | 3, 4 | 448 | ||
GenBank mRNA | AK300003.1 | - | 3, 4 | 427 | ||
AK311297.1 | - | 2, 3 | 473 | |||
BC111483.1 | - | 3, 4 | 421 | 3877097 | ||
JX960761.1 | - | 3, 4 | 376 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
