siRNA Details
Gene Symbol: LOC344887 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033752.2 | 4 | 624 | |||
GenBank mRNA | AK309314.1 | - | 4 | 370 | ||
AK309325.1 | - | 4 | 624 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
