siRNA Details
Gene Symbol: DNAH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001370.1 | NP_001361.1 | 30 | 4762 | ||
XM_006711956.2 | XP_006712019.1 | 30 | 4867 | |||
XM_011532649.2 | XP_011530951.1 | 30 | 4716 | |||
XM_011532650.2 | XP_011530952.1 | 30 | 4717 | |||
XM_011532652.1 | XP_011530954.1 | 29 | 4636 | |||
XM_011532653.1 | XP_011530955.1 | 30 | 4762 | |||
XM_011532654.1 | XP_011530956.1 | 29 | 4495 | |||
XM_011532655.2 | XP_011530957.1 | 26 | 3784 | |||
XM_011532657.1 | XP_011530959.1 | 24 | 3523 | |||
XM_011532660.1 | XP_011530962.1 | 30 | 4762 | |||
XM_017003521.1 | XP_016859010.1 | 29 | 4636 | |||
XM_017003522.1 | XP_016859011.1 | 25 | 4588 | |||
XM_017003523.1 | XP_016859012.1 | 25 | 3755 | |||
XM_017003524.1 | XP_016859013.1 | 30 | 4762 | |||
GenBank mRNA | AJ132086.1 | - | 2 | 200 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
