siRNA Details
Gene Symbol: DNAH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001370.1 | NP_001361.1 | 30, 31 | 4788 | ||
XM_006711956.2 | XP_006712019.1 | 30, 31 | 4893 | |||
XM_011532649.2 | XP_011530951.1 | 30, 31 | 4742 | |||
XM_011532650.2 | XP_011530952.1 | 30, 31 | 4743 | |||
XM_011532652.1 | XP_011530954.1 | 29, 30 | 4662 | |||
XM_011532653.1 | XP_011530955.1 | 30, 31 | 4788 | |||
XM_011532654.1 | XP_011530956.1 | 29, 30 | 4521 | |||
XM_011532655.2 | XP_011530957.1 | 26, 27 | 3810 | |||
XM_011532657.1 | XP_011530959.1 | 24, 25 | 3549 | |||
XM_011532660.1 | XP_011530962.1 | 30, 31 | 4788 | |||
XM_017003521.1 | XP_016859010.1 | 29, 30 | 4662 | |||
XM_017003522.1 | XP_016859011.1 | 25, 26 | 4614 | |||
XM_017003523.1 | XP_016859012.1 | 25, 26 | 3781 | |||
XM_017003524.1 | XP_016859013.1 | 30, 31 | 4788 | |||
GenBank mRNA | AJ132086.1 | - | 2, 3 | 226 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
