siRNA Details
Gene Symbol: ACTBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017992.3 | NP_001017992.1 | 1 | 694 | ||
GenBank mRNA | BC137470.1 | - | 1 | 612 | 9021470 | |
BC137473.1 | - | 1 | 612 | 9021473 | ||
BX648504.1 | - | 1 | 695 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
