siRNA Details
Gene Symbol: RNF150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020724.1 | NP_065775.1 | 7 | 2637 | ||
XM_005263150.4 | XP_005263207.1 | 8 | 2151 | |||
XM_011532147.2 | XP_011530449.1 | 8 | 1614 | |||
XM_011532148.2 | XP_011530450.1 | 8 | 1629 | |||
XM_017008475.1 | XP_016863964.1 | 7 | 1548 | |||
XM_017008476.1 | XP_016863965.1 | 7 | 1521 | |||
GenBank mRNA | AB033040.1 | - | 3087 | |||
AK130520.1 | - | 8 | 1649 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
