siRNA Details
Gene Symbol: FAM83B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010872.2 | NP_001010872.1 | 5 | 994 | ||
XM_006715022.3 | XP_006715085.1 | 5 | 1023 | |||
XM_011514394.2 | XP_011512696.1 | 5 | 1036 | |||
XM_011514395.2 | XP_011512697.1 | 5 | 2025 | |||
XM_017010478.1 | XP_016865967.1 | 5 | 1020 | |||
GenBank mRNA | AK055204.1 | - | 5 | 995 | ||
BC101628.1 | - | 4 | 953 | 8069134 | ||
BC112275.1 | - | 4 | 953 | 8327743 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
