siRNA Details
Gene Symbol: ZNF131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297548.1 | NP_001284477.1 | 5 | 769 | ||
NR_123720.1 | 4 | 673 | ||||
XM_005248359.2 | XP_005248416.1 | 5 | 984 | |||
XM_005248360.2 | XP_005248417.1 | 5 | 781 | |||
XM_005248361.2 | XP_005248418.1 | 5 | 1009 | |||
XM_005248362.4 | XP_005248419.1 | 5 | 958 | |||
XM_005248363.4 | XP_005248420.1 | 5 | 954 | |||
XM_005248367.2 | XP_005248424.1 | 4 | 807 | |||
XM_011514123.2 | XP_011512425.1 | 3 | 651 | |||
XM_011514124.2 | XP_011512426.1 | 4 | 666 | |||
XM_017009830.1 | XP_016865319.1 | 5 | 774 | |||
XM_017009831.1 | XP_016865320.1 | 3 | 943 | |||
XM_017009839.1 | XP_016865328.1 | 4 | 890 | |||
XM_017009840.1 | XP_016865329.1 | 4 | 646 | |||
GenBank mRNA | AK299315.1 | - | 6 | 769 | ||
AK307977.1 | - | 4 | 631 | |||
U09410.1 | - | 1 | 289 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: AY092766, AK135723, BC057991, AK047429, AK162903.
Gene Ontology Categories:
