siRNA Details
Gene Symbol: ZNF131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297548.1 | NP_001284477.1 | 5 | 812 | ||
NR_123720.1 | 4 | 716 | ||||
XM_005248359.2 | XP_005248416.1 | 5 | 1027 | |||
XM_005248360.2 | XP_005248417.1 | 5 | 824 | |||
XM_005248361.2 | XP_005248418.1 | 5 | 1052 | |||
XM_005248362.4 | XP_005248419.1 | 5 | 1001 | |||
XM_005248363.4 | XP_005248420.1 | 5 | 997 | |||
XM_005248367.2 | XP_005248424.1 | 4 | 850 | |||
XM_011514123.2 | XP_011512425.1 | 3 | 694 | |||
XM_011514124.2 | XP_011512426.1 | 4 | 709 | |||
XM_017009830.1 | XP_016865319.1 | 5 | 817 | |||
XM_017009831.1 | XP_016865320.1 | 3 | 986 | |||
XM_017009839.1 | XP_016865328.1 | 4 | 933 | |||
XM_017009840.1 | XP_016865329.1 | 4 | 689 | |||
GenBank mRNA | AK299315.1 | - | 6 | 812 | ||
AK307977.1 | - | 4 | 674 | |||
U09410.1 | - | 1 | 332 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
