siRNA Details
Gene Symbol: PWWP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130864.1 | NP_001124336.1 | 2 | 1436 | ||
NM_001267035.1 | NP_001253964.1 | 2 | 1436 | |||
NM_052927.2 | NP_443159.1 | 2 | 1436 | |||
XM_005265811.3 | XP_005265868.1 | 2 | 911 | |||
XM_005265812.2 | XP_005265869.1 | 3 | 1533 | |||
XM_006714817.3 | XP_006714880.1 | 4 | 2818 | |||
XM_011534424.2 | XP_011532726.1 | 2 | 1476 | |||
XM_017009009.1 | XP_016864498.1 | 4 | 2392 | |||
XM_017009010.1 | XP_016864499.1 | 3 | 2628 | |||
XM_017009011.1 | XP_016864500.1 | 4 | 1110 | |||
XM_017009012.1 | XP_016864501.1 | 2 | 1475 | |||
GenBank mRNA | AK055921.1 | - | 1 | 661 | ||
BC035076.2 | - | 2 | 1414 | |||
BC105110.1 | - | 2 | 1413 | 8144113 | ||
BC143437.1 | - | 2 | 1411 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
