siRNA Details
Gene Symbol: SMIM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001124767.1 | NP_001118239.1 | 2 | 315 | ||
XM_011533729.2 | XP_011532031.1 | 2 | 431 | |||
GenBank mRNA | AK095910.1 | - | 2 | 956 | ||
BC017996.1 | - | 2 | 315 | |||
BG031013.1 | - | 2 | 305 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
