siRNA Details
Gene Symbol: SYNPO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128933.2 | NP_001122405.1 | 4 | 3653 | ||
GenBank mRNA | AF177291.1 | - | 1 | 578 | ||
AJ010482.1 | - | 2 | 2673 | |||
AK296962.1 | - | 2 | 2419 | |||
AL832031.1 | - | 4 | 3636 | |||
AL832363.1 | - | 4 | 3547 | |||
AL833294.1 | - | 4 | 3639 | |||
BX647849.1 | - | 2 | 3162 | |||
FN422000.1 | - | 2 | 2708 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
