siRNA Details
Gene Symbol: LURAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013615.2 | NP_001013633.1 | 2 | 811 | ||
GenBank mRNA | AK057892.1 | - | 2 | 811 | ||
BC034422.1 | - | 2 | 804 | 5163612 | ||
BX370151.2 | - | 1 | 131 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
