siRNA Details
Gene Symbol: LURAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013615.2 | NP_001013633.1 | 2 | 1727 | ||
GenBank mRNA | AK057892.1 | - | 2 | 1727 | ||
CR746264.1 | - | 1 | 288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
