siRNA Details
Gene Symbol: TMEM223 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080501.2 | NP_001073970.1 | 2 | 356 | ||
GenBank mRNA | BC008643.1 | - | 2 | 334 | ||
BC020914.1 | - | 2 | 350 | |||
BC042801.1 | - | 2 | 294 | |||
BC094837.1 | - | 2 | 334 | |||
BC127679.1 | - | 2 | 363 | 40127503 | ||
BP872883.1 | - | 2 | 356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
