siRNA Details
Gene Symbol: LOC283922 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026950.1 | 8 | 1187 | |||
GenBank mRNA | AK096768.1 | - | 8 | 1172 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
