siRNA Details
Gene Symbol: LOC101927506 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017014131.1 | XP_016869620.1 | 3 | 2156 | ||
XM_017014132.1 | XP_016869621.1 | 3 | 2156 | |||
XM_017014133.1 | XP_016869622.1 | 3 | 2156 |
Gene Symbol: LOC728323 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130702.1 | 4 | 473 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
