siRNA Details
Gene Symbol: FAM161A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201543.1 | NP_001188472.1 | 3 | 891 | ||
NM_032180.2 | NP_115556.2 | 3 | 891 | |||
NR_037710.1 | 3 | 835 | ||||
XM_017005072.1 | XP_016860561.1 | 3 | 844 | |||
XM_017005073.1 | XP_016860562.1 | 3 | 818 | |||
XM_017005074.1 | XP_016860563.1 | 3 | 791 | |||
GenBank mRNA | AK023367.1 | - | 4 | 1050 | ||
AK296255.1 | - | 2 | 425 | |||
AK310908.1 | - | 3 | 776 | |||
BC036448.1 | - | 3 | 789 | |||
BC107162.1 | - | 3 | 663 | |||
BC107163.1 | - | 3 | 663 | |||
BX648834.1 | - | 3 | 801 | |||
BX649029.1 | - | 3 | 802 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
