siRNA Details
Gene Symbol: FAM161A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201543.1 | NP_001188472.1 | 5 | 1887 | ||
NM_032180.2 | NP_115556.2 | 4 | 1719 | |||
NR_037710.1 | 4 | 1663 | ||||
XM_017005072.1 | XP_016860561.1 | 4 | 1672 | |||
XM_017005073.1 | XP_016860562.1 | 5 | 1814 | |||
XM_017005074.1 | XP_016860563.1 | 4 | 1619 | |||
GenBank mRNA | AK023367.1 | - | 5 | 1878 | ||
AK296255.1 | - | 3 | 1253 | |||
AK310908.1 | - | 6 | 2062 | |||
BC036448.1 | - | 4 | 1617 | |||
BC107162.1 | - | 4 | 1491 | |||
BC107163.1 | - | 4 | 1491 | |||
BX648834.1 | - | 5 | 1797 | |||
BX649029.1 | - | 4 | 1630 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
