siRNA Details
Gene Symbol: MURC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018116.2 | NP_001018126.1 | 2 | 1150 | ||
GenBank mRNA | AK300099.1 | - | 2 | 1150 | ||
BC090888.1 | - | 2 | 1067 | 30529833 | ||
EU487253.1 | - | 2 | 1085 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
