siRNA Details
Gene Symbol: CARNS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166222.1 | NP_001159694.1 | 10 | 3120 | ||
NM_020811.1 | NP_065862.1 | 9 | 3093 | |||
XM_011545189.2 | XP_011543491.1 | 9 | 3410 | |||
XM_011545191.2 | XP_011543493.1 | 8 | 5768 | |||
XM_017018054.1 | XP_016873543.1 | 9 | 4324 | |||
XM_017018055.1 | XP_016873544.1 | 7 | 3604 | |||
GenBank mRNA | AB037815.1 | - | 8 | 4218 | ||
BC036557.1 | - | 1 | 1491 | |||
EF560746.1 | - | 9 | 3098 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
