siRNA Details
Gene Symbol: CARNS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011545191.2 | XP_011543493.1 | 1 | 1558 | ||
XM_017018054.1 | XP_016873543.1 | 2 | 1260 | |||
XM_017018055.1 | XP_016873544.1 | 1 | 630 | |||
GenBank mRNA | AB037815.1 | - | 1 | 1154 | ||
AK295331.1 | - | 2 | 1260 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
