siRNA Details
Gene Symbol: GRIFIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291784.1 | NP_001278713.1 | 2 | 112 | ||
XM_011515405.2 | XP_011513707.1 | 3 | 292 | |||
GenBank mRNA | CN281639.1 | - | 3 | 271 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
