siRNA Details
Gene Symbol: LOC339803 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036496.1 | 2 | 1996 | |||
GenBank mRNA | AK023856.1 | - | 2 | 1970 | ||
BC008254.1 | - | 1 | 491 | 3872056 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
