siRNA Details
Gene Symbol: C1orf100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012970.2 | NP_001012988.1 | 4 | 328 | ||
XM_011544124.2 | XP_011542426.1 | 5 | 1170 | |||
XM_011544125.2 | XP_011542427.1 | 4 | 920 | |||
XM_011544128.2 | XP_011542430.1 | 4 | 1431 | |||
XM_017000571.1 | XP_016856060.1 | 5 | 1200 | |||
XM_017000572.1 | XP_016856061.1 | 5 | 1708 | |||
GenBank mRNA | HY055397.1 | - | 4 | 284 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
