siRNA Details
Gene Symbol: C1orf100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012970.2 | NP_001012988.1 | 2 | 137 | ||
NM_001276348.1 | NP_001263277.1 | 2 | 137 | |||
NM_001276349.1 | NP_001263278.1 | 2 | 137 | |||
XM_011544124.2 | XP_011542426.1 | 3 | 979 | |||
XM_011544125.2 | XP_011542427.1 | 2 | 705 | |||
XM_011544128.2 | XP_011542430.1 | 2 | 1216 | |||
XM_011544129.2 | XP_011542431.1 | 3 | 986 | |||
XM_011544130.2 | XP_011542432.1 | 3 | 984 | |||
XM_017000571.1 | XP_016856060.1 | 3 | 985 | |||
XM_017000572.1 | XP_016856061.1 | 3 | 1493 | |||
GenBank mRNA | BC054479.1 | - | 2 | 141 | ||
BX099227.1 | - | 2 | 123 | |||
HY055397.1 | - | 2 | 93 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
