siRNA Details
Gene Symbol: GALNTL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034845.2 | NP_001030017.2 | 7 | 1604 | ||
XM_011531993.2 | XP_011530295.1 | 6 | 942 | |||
XM_011531994.1 | XP_011530296.1 | 6 | 776 | |||
XM_011531995.1 | XP_011530297.1 | 5 | 687 | |||
XM_011531996.1 | XP_011530298.1 | 4 | 587 | |||
XM_011531997.1 | XP_011530299.1 | 4 | 509 | |||
XM_017008243.1 | XP_016863732.1 | 7 | 2338 | |||
XM_017008244.1 | XP_016863733.1 | 6 | 1710 | |||
GenBank mRNA | AJ626725.1 | - | 6 | 817 | ||
BC047551.1 | - | 7 | 1525 | 5301536 | ||
GU220060.1 | - | 6 | 868 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_175032, GU220061.
Gene Ontology Categories:
