siRNA Details
Gene Symbol: GALNTL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034845.2 | NP_001030017.2 | 9 | 1854 | ||
XM_011531993.2 | XP_011530295.1 | 8 | 1192 | |||
XM_011531994.1 | XP_011530296.1 | 8 | 1026 | |||
XM_011531995.1 | XP_011530297.1 | 7 | 937 | |||
XM_011531996.1 | XP_011530298.1 | 6 | 837 | |||
XM_011531997.1 | XP_011530299.1 | 6 | 759 | |||
XM_017008243.1 | XP_016863732.1 | 9 | 2588 | |||
XM_017008244.1 | XP_016863733.1 | 8 | 1960 | |||
GenBank mRNA | AJ626725.1 | - | 8 | 1067 | ||
BC047551.1 | - | 9 | 1775 | 5301536 | ||
GU220060.1 | - | 8 | 1118 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
