siRNA Details
Gene Symbol: HES3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024598.3 | NP_001019769.1 | 2 | 136 | ||
GenBank mRNA | CN370501.1 | - | 2 | 152 | ||
CN413592.1 | - | 2 | 128 | |||
CX755241.1 | - | 2 | 134 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
