siRNA Details
Gene Symbol: AIM1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039775.3 | NP_001034864.2 | 13 | 4211 | ||
XM_005245918.2 | XP_005245975.1 | 13 | 4197 | |||
XM_011541672.1 | XP_011539974.1 | 12 | 4161 | |||
XM_011541673.2 | XP_011539975.1 | 13 | 4436 | |||
GenBank mRNA | AK095339.1 | - | 12 | 1255 | ||
BC136870.1 | - | 9 | 953 | 9020861 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
