siRNA Details
Gene Symbol: GPR149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038705.1 | NP_001033794.1 | 3 | 1578 | ||
GenBank mRNA | AY255534.1 | - | 3 | 1013 | ||
DQ318360.1 | - | 3 | 1578 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
