siRNA Details
Gene Symbol: CCDC192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317938.1 | NP_001304867.1 | 3 | 241 | ||
XM_017009805.1 | XP_016865294.1 | 2 | 303 | |||
XM_017009806.1 | XP_016865295.1 | 3 | 207 | |||
XM_017009807.1 | XP_016865296.1 | 3 | 198 | |||
XM_017009808.1 | XP_016865297.1 | 2 | 129 | |||
XM_017009809.1 | XP_016865298.1 | 3 | 272 | |||
XM_017009810.1 | XP_016865299.1 | 3 | 271 | |||
XM_017009811.1 | XP_016865300.1 | 3 | 271 | |||
XM_017009812.1 | XP_016865301.1 | 3 | 272 | |||
XM_017009813.1 | XP_016865302.1 | 3 | 270 | |||
GenBank mRNA | HY052603.1 | - | 3 | 209 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
